JP
EN
Applications
Products
Services
Support
Home
Search Results
TWEEN
®
20, PROTEIN GRADE
®
Detergent, 10% Solution, Sterile-Filtered
(1)
TWEEN
®
20, PROTEIN GRADE
®
Detergent, 10% Solution, Sterile-Filtered
Synonyms:
TWEEN
®
20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
CAS Number:
9005-64-5
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
655206
TWEEN
®
20, PROTEIN GRADE
®
Detergent, 10% Solution, Sterile-Filtered, Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins. Conductivity:50 µS/cm.
Expand
View Pricing