Merck
  • Clinical and Genomic Correlates of Neutrophil Reactive Oxygen Species Production in Pediatric Patients With Crohn's Disease.
Clinical and Genomic Correlates of Neutrophil Reactive Oxygen Species Production in Pediatric Patients With Crohn's Disease.
Gastroenterology (2018-02-20)
Lee A Denson, Ingrid Jurickova, Rebekah Karns, Kelly A Shaw, David J Cutler, David T Okou, Anne Dodd, Kathryn Quinn, Kajari Mondal, Bruce J Aronow, Yael Haberman, Aaron Linn, Adam Price, Ramona Bezold, Kathleen Lake, Kimberly Jackson, Thomas D Walters, Anne Griffiths, Robert N Baldassano, Joshua D Noe, Jeffrey S Hyams, Wallace V Crandall, Barbara S Kirschner, Melvin B Heyman, Scott Snapper, Stephen L Guthery, Marla C Dubinsky, Neal S Leleiko, Anthony R Otley, Ramnik J Xavier, Christine Stevens, Mark J Daly, Michael E Zwick, Subra Kugathasan
ABSTRACT
Individuals with monogenic disorders of phagocyte function develop chronic colitis that resembles Crohn's disease (CD). We tested for associations between mutations in genes encoding reduced nicotinamide adenine dinucleotide phosphate (NADPH) oxidases, neutrophil function, and phenotypes of CD in pediatric patients. We performed whole-exome sequence analysis to identify mutations in genes encoding NADPH oxidases (such as CYBA, CYBB, NCF1, NCF2, NCF4, RAC1, and RAC2) using DNA from 543 pediatric patients with inflammatory bowel diseases. Blood samples were collected from an additional 129 pediatric patients with CD and 26 children without IBD (controls); we performed assays for neutrophil activation, reactive oxygen species (ROS) production, and bacteria uptake and killing. Whole-exome sequence analysis was performed using DNA from 46 of the children with CD to examine associations with NADPH gene mutations; RNA sequence analyses were performed using blood cells from 46 children with CD to test for variations in neutrophil gene expression associated with ROS production. We identified 26 missense mutations in CYBA, CYBB, NCF1, NCF2, and NCF4. Patients with CD who carried mutations in these genes were 3-fold more likely to have perianal disease (P = .0008) and stricturing complications (P = .002) than children with CD without these mutations. Among patients with CD with none of these mutations, 9% had undergone abdominal surgery; among patients with mutations in these NADPH oxidase genes, 31% had undergone abdominal surgery (P = .0004). A higher proportion of neutrophils from children with CD had low ROS production (47%) than from controls (15%) among the 129 patients tested for ROS (P = .002). Minor alleles of the NADPH genes were detected in 7% of children with CD whose neutrophils produced normal levels of ROS vs 38% of children whose neutrophils produced low levels of ROS (P = .009). Neutrophils that produced low levels of ROS had specific alterations in genes that regulate glucose metabolism and antimicrobial responses. We identified missense mutations in genes that encode NADPH oxidases in children with CD; these were associated with a more aggressive disease course and reduced ROS production by neutrophils from the patients.
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Pricing and availability is not currently available.