JP
EN
Applications
Products
Services
Support
Home
Search Results
Ovum pick up and in vitro embryo production in cows superstimulated with an individually adapted superstimulation protocol
Ovum pick up and in vitro embryo production in cows superstimulated with an individually adapted superstimulation protocol
Animal Reproduction Science (2005)
De R, et al.