  • Search Results
  • Study on Biological Activity of Bread Enriched with Natural Polyphenols in Terms of Growth Inhibition of Tumor Intestine Cells.
Study on Biological Activity of Bread Enriched with Natural Polyphenols in Terms of Growth Inhibition of Tumor Intestine Cells.
Journal of medicinal food (2019-09-11)
Dominik Szwajgier, Roman Paduch, Wirginia Kukuła-Koch, Magdalena Polak-Berecka, Adam Waśko
ABSTRACT
A complex plant polyphenolic preparation (PP) was produced from chokeberry, raspberry, wild strawberry, peach, bilberry, apricot, cranberry, and parsley, using ultrafiltration and C18 preparative chromatography. Thirty main compounds were identified in PP (LC-MS), with the highest contribution of cyanidin 3-O-glucoside, p-coumaroyl glucoside, chlorogenic acid, neochlorogenic acid, and isoquercetin. PP was used (at 0.16% m/m) for the production of a sourdough bread (based on rye flour, water, and salt), followed by in vitro digestion. Fluid obtained after PP-enriched bread digestion (EBD fluid) was tested in terms of cytotoxicity, growth inhibition, antioxidant activity, and morphological changes in cancerous intestinal epithelial cells (HT-29) and normal (CCD 841 CoTr). Results show that EBD fluid concentration over 125 μg/mL significantly decreased activity of succinate dehydrogenase in HT-29 cells and reduced their viability of 25%. At this concentration of EBD fluid, modification in cellular morphology was also observed. DPPH analysis revealed that the highest antioxidant activity was observed at concentration of 75 μg/mL, both PP and EBD fluid. Our results show that an introduction of PP into relatively low-polyphenolic, baking products should be carefully considered because polyphenols still retain its biological activity. Antioxidant activity of polyphenols is one of the mechanisms that explains the observed effect of inhibiting the growth of colon cancer cells.
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
(+)-Arabinogalactan, from larch wood
Sigma-Aldrich
Menadione, crystalline
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, ≥98.0%
Sigma-Aldrich
Amylopectin from maize
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Hemin, from bovine, ≥90%