  • Search Results
  • Protein Arginine Methyltransferase 1 Interacts With PGC1α and Modulates Thermogenic Fat Activation.
Protein Arginine Methyltransferase 1 Interacts With PGC1α and Modulates Thermogenic Fat Activation.
Endocrinology (2019-09-27)
Xiaona Qiao, Dong-Il Kim, Heejin Jun, Yingxu Ma, Alexander J Knights, Min-Jung Park, Kezhou Zhu, Jay H Lipinski, Jiling Liao, Yiming Li, Stéphane Richard, Steven A Weinman, Jun Wu
ABSTRACT
Protein arginine methyltransferases (PRMTs) are enzymes that regulate the evolutionarily conserved process of arginine methylation. It has been reported that PRMTs are involved in many metabolic regulatory pathways. However, until now, their roles in adipocyte function, especially browning and thermogenesis, have not been evaluated. Even though Prmt1 adipocyte-specific-deleted mice (Prmt1fl/flAQcre) appeared normal at basal level, following cold exposure or β-adrenergic stimulation, impaired induction of the thermogenic program was observed in both the interscapular brown adipose tissue and inguinal white adipose tissue of Prmt1fl/flAQcre mice compared with littermate controls. Different splicing variants of Prmt1 have been reported. Among them, PRMT1 variant 1 and PRMT1 variant 2 (PRMT1V2) are well conserved between humans and mice. Both variants contribute to the activation of thermogenic fat, with PRMT1V2 playing a more dominant role. Mechanistic studies using cultured murine and human adipocytes revealed that PRMT1V2 mediates thermogenic fat activation through PGC1α, a transcriptional coactivator that has been shown to play a key role in mitochondrial biogenesis. To our knowledge, our data are the first to demonstrate that PRMT1 plays a regulatory role in thermogenic fat function. These findings suggest that modulating PRMT1 activity may represent new avenues to regulate thermogenic fat and mediate energy homeostasis. This function is conserved in human primary adipocytes, suggesting that further investigation of this pathway may ultimately lead to therapeutic strategies against human obesity and associated metabolic disorders.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Newborn Calf Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Anti-PGC-1α Rabbit pAb, liquid, Calbiochem®