Merck
  • Proteomic analysis of defined HDL subpopulations reveals particle-specific protein clusters: relevance to antioxidative function.
Proteomic analysis of defined HDL subpopulations reveals particle-specific protein clusters: relevance to antioxidative function.
Arteriosclerosis, thrombosis, and vascular biology (2009-03-28)
W Sean Davidson, R A Gangani D Silva, Sandrine Chantepie, William R Lagor, M John Chapman, Anatol Kontush
ABSTRACT
Recent proteomic studies have identified multiple proteins that coisolate with human HDL. We hypothesized that distinct clusters of protein components may distinguish between physicochemically-defined subpopulations of HDL particles, and that such clusters may exert specific biological function(s). We investigated the distribution of proteins across 5 physicochemically-defined particle subpopulations of normolipidemic human HDL (HDL2b, 2a, 3a, 3b, 3c) fractionated by isopycnic density gradient ultracentrifugation. Liquid chromatography/electrospray mass spectrometry identified a total of 28 distinct HDL-associated proteins. Using an abundance pattern analysis of peptide counts across the HDL subfractions, these proteins could be grouped into 5 distinct classes. A more in-depth correlational network analysis suggested the existence of distinct protein clusters, particularly in the dense HDL3 particles. Levels of specific HDL proteins, primarily apoL-I, PON1, and PON3, correlated with the potent capacity of HDL3 to protect LDL from oxidation. These findings suggest that HDL is composed of distinct particles containing unique (apolipo)protein complements. Such subspeciation forms a potential basis for understanding the numerous observed functions of HDL. Further work using additional separation techniques will be required to define these species in more detail.
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard