  • Search Results
  • TC-E 5003, a protein methyltransferase 1 inhibitor, activates the PKA-dependent thermogenic pathway in primary murine and human subcutaneous adipocytes.
TC-E 5003, a protein methyltransferase 1 inhibitor, activates the PKA-dependent thermogenic pathway in primary murine and human subcutaneous adipocytes.
FEBS letters (2020-08-09)
Min-Jung Park, Jiling Liao, Dong-Il Kim
ABSTRACT
We previously reported the involvement of protein arginine methyltransferase 1 (PRMT1) in adipocyte thermogenesis. Here, we investigate the effects of PRMT1 inhibitors on thermogenesis. Unexpectedly, we find that the PRMT1 inhibitor TC-E 5003 (TC-E) induces the thermogenic properties of primary murine and human subcutaneous adipocytes. TC-E treatment upregulates the expression of Ucp1 and Fgf21 significantly and activates protein kinase A signaling and lipolysis in primary subcutaneous adipocytes from both mouse and humans. We further find that the thermogenic effects of TC-E are independent of PRMT1 and beta-adrenergic receptors. Our data indicate that TC-E exerts strong effects on murine and human subcutaneous adipocytes by activating beige adipocytes via PKA signaling.
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Furamidine dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)