Merck
  • v-cbl, an oncogene from a dual-recombinant murine retrovirus that induces early B-lineage lymphomas.
v-cbl, an oncogene from a dual-recombinant murine retrovirus that induces early B-lineage lymphomas.
Proceedings of the National Academy of Sciences of the United States of America (1989-02-01)
W Y Langdon, J W Hartley, S P Klinken, S K Ruscetti, H C Morse
ABSTRACT
Cas NS-1 is an acutely transforming murine retrovirus that induces pre-B and pro-B cell lymphomas. Molecular cloning showed it was generated from the ecotropic Cas-Br-M virus by sequential recombinations with endogenous retroviral sequences and a cellular oncogene. The oncogene sequence shows no homology with known oncogenes but some similarity to the yeast transcriptional activator GCN4. A 100-kDa gag-cbl fusion protein, with no detectable kinase activity, is responsible for the cellular transformation. The cellular homologue of v-cbl, present in mouse and human DNA, is expressed in a range of hemopoietic lineages.