  • Search Results
  • Characterization of backbone dynamics using solution NMR spectroscopy to discern the functional plasticity of structurally analogous proteins.
Characterization of backbone dynamics using solution NMR spectroscopy to discern the functional plasticity of structurally analogous proteins.
STAR protocols (2021-11-12)
Ashish A Kawale, Björn M Burmann
ABSTRACT
The comprehensive delineation of inherent dynamic motions embedded in proteins, which can be crucial for their functional repertoire, is often essential yet remains poorly understood in the majority of cases. In this protocol, we outline detailed descriptions of the necessary steps for employing solution NMR spectroscopy for the in-depth amino acid level understanding of backbone dynamics of proteins. We describe the application of the protocol on the structurally analogous Tudor domains with disparate functionalities as a model system. For complete details on the use and execution of this protocol, please refer to Kawale and Burmann (2021).
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Ammonium-15N chloride, ≥98 atom % 15N, ≥99% (CP)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, ≥98.0%
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Pyridoxal hydrochloride, ≥99.5% (T)