Merck
  • Photoinduced electron transfer reaction in polymer-surfactant aggregates: Photoinduced electron transfer between N,N-dimethylaniline and 7-amino coumarin dyes.
Photoinduced electron transfer reaction in polymer-surfactant aggregates: Photoinduced electron transfer between N,N-dimethylaniline and 7-amino coumarin dyes.
The Journal of chemical physics (2008-06-03)
Anjan Chakraborty, Debabrata Seth, Palash Setua, Nilmoni Sarkar
ABSTRACT
Photoinduced electron transfer between coumarin dyes and N,N-dimethylaniline has been investigated by using steady state and picosecond time resolved fluorescence spectroscopy in sodium dodecyl sulphate (SDS) micelles and PVP-polyvinyl pyrrolidone (SDS) polymer-surfactant aggregates. A slower rate of electron transfer is observed in PVP-SDS aggregates than in polymer-free SDS micelles. A Marcus type inversion is observed in the correlation of free energy change in comparison with the electron transfer rate. The careful investigation reveals that C-151 deviates from the normal Marcus inverted region compared to its analogs C-152 and C-481 due to slower rotational relaxation and smaller translational diffusion coefficient.
Sigma-Aldrich
N,N-Dimethylaniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
Sigma-Aldrich
N,N-Dimethylaniline, ReagentPlus®, 99%