Merck
  • Templated synthesis of a large and flexible covalent porphyrinic cage bearing orthogonal recognition sites.
Templated synthesis of a large and flexible covalent porphyrinic cage bearing orthogonal recognition sites.
Chemical communications (Cambridge, England) (2012-04-21)
Julien Taesch, Valérie Heitz, Filip Topić, Kari Rissanen
ABSTRACT
A large covalent cage incorporating two porphyrins attached by four long and flexible polyether chains each bearing two 3-pyridyl ligands was synthesized from a DABCO-templated olefin metathesis reaction. The X-ray structure of the cage with the DABCO coordinated inside the cavity to the two zinc(II) porphyrins reveals a highly symmetric structure.
Sigma-Aldrich
Dabco® 33-LV
Pricing and availability is not currently available.
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Pricing and availability is not currently available.