Merck
  • Isolation and characterization of unusual hydrazides from Streptomyces sp. impact of the cultivation support and extraction procedure.
Isolation and characterization of unusual hydrazides from Streptomyces sp. impact of the cultivation support and extraction procedure.
Journal of natural products (2013-02-08)
Géraldine Le Goff, Marie-Thérèse Martin, Bogdan I Iorga, Emilie Adelin, Claudine Servy, Sylvie Cortial, Jamal Ouazzani
ABSTRACT
Three novel hydrazides, geralcins C-E (1-3), were isolated from Streptomyces sp. LMA-545, together with MH-031 and geralcins A and B. This unusual family of compounds was isolated from liquid-state and agar-supported fermentation using Amberlite XAD-16 solid-phase extraction during the cultivation step. The use of such neutral resin during the cultivation step allowed the specific adsorption of microbial secondary metabolites, avoiding any contamination of the crude extracts by the constituents of the culture medium. The trapped compounds were eluted from the resin with methanol, and their structures elucidated using (1)H, (13)C, and (15)N NMR spectroscopic analysis and high-resolution mass spectrometry. Molecular modeling calculations were applied in order to support structural attributions. No antimicrobial, cytotoxic, or DnaG-inhibition activities were detected for geralcins D and E. Geralcin C has no antimicrobial activity but exhibited an IC(50) of 0.8 μM against KB and HCT116 cancer cell lines. Furthermore, geralcin C inhibited the E. coli DnaG primase, a Gram-negative antimicrobial target, with an IC(50) of 0.7 mM.
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
γ-Butyrolactone, SAJ first grade, ≥99.5%