Merck
  • Prevalence, morphology, and natural history of FGFR1-amplified lung cancer, including squamous cell carcinoma, detected by FISH and SISH.
Prevalence, morphology, and natural history of FGFR1-amplified lung cancer, including squamous cell carcinoma, detected by FISH and SISH.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2014-04-26)
Prudence A Russell, Yong Yu, Richard J Young, Matthew Conron, Zoe Wainer, Naveed Alam, Benjamin Solomon, Gavin M Wright
ABSTRACT
The aim of this study was to investigate the prevalence of fibroblast growth factor receptor 1 (FGFR1) amplification by fluorescence in situ hybridization (FISH) in a lung cancer patient cohort and to correlate results with morphology, silver in situ hybridization (SISH), and patient outcome. FGFR1 FISH and SISH were performed in 406 and 385 lung cancer cases, respectively, and the results were compared. High-level FGFR1 amplification was defined as the ratio of FGFR1/centromere 8 ≥2, or tumor cell percentage with ≥15 signals ≥10%, or average number of signals/tumor cell nucleus ≥6. Low-level amplification was defined as tumor cell percentage with ≥5 signals ≥50%. Of 406 tumors tested, there were 191 squamous cell carcinomas, 28 carcinomas with focal squamous morphology, 24 large cell carcinomas with squamous immunoprofile, 115 adenocarcinomas, 17 neuroendocrine tumors, and 31 carcinomas without squamous morphology or immunoprofile. FGFR1 FISH was assessable in 368 tumors, with FGFR1 amplification identified in 50, including 48 tumors with either squamous morphology or immunoprofile (48 of 225, 21.3%), and two 'marker-null' tumors without squamous or glandular morphology or immunoprofile (2 of 143, 1.4%; P<0.0001). FGFR1 SISH was assessable in 347 tumors. All 46 FGFR1 FISH-amplified tumors with tumor available for testing showed amplification with SISH, while all other tumors were negative. There was no relationship between FGFR1 amplification status and disease-free (P=0.88, HR=1.04, 95% confidence interval (CI)=0.67-1.60) or overall survival (P=0.97, HR=1.01, 95% CI=0.65-1.58) in surgically radically treated patients with tumors with any squamous morphology or immunoprofile. FGFR1 amplification is a common abnormality in tumors with any squamous morphology or immunoprofile, but it is also present in 'marker-null' tumors. The results of FGFR1 SISH showed 1:1 correlation with the results of FGFR1 FISH, indicating that SISH may be an alternative method to detect FGFR1 amplification. No relationship was detected between patient outcome and FGFR1 amplification.
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dinitrophenol, SAJ special grade, ≥98.0%
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin