Merck
Controllable coating of microneedles for transdermal drug delivery.
Drug development and industrial pharmacy (2014-01-01)
Jianmin Chen, Yuqin Qiu, Suohui Zhang, Guozhong Yang, Yunhua Gao
ABSTRACT
Coated microneedles have been paid much attention recently, and several coating strategies have been developed to address the problems during coating process. However, there are still some unresolved issues, such as, precise control requirements, microneedle substrate contamination and high processing temperature. The purpose of this study was to develop a simple and controllable method to make uniform coatings on microneedles at room temperature. This novel method avoids the contamination of microneedle substrate by providing both the adsorption force of thickener and micro-scale coating film produced by a newly design device. Thickeners were screened to enhance the mass of coatings. The parameters that influence the coatings were tested systematically, which made coating process controllable. Finally, three model drugs were coated onto microneedles to prove the method is applicable more broadly. In addition, insertion experiments were carried out to test the drug delivery feasibility of the coated microneedles. In conclusion, this study presents a simple and controllable method to coat microneedles with small molecular chemical drugs or large proteins for rapid skin drug delivery.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt, tested according to Ph. Eur.
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Riboflavin sodium phosphate, European Pharmacopoeia (EP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard