- Increased efficacy of metformin corresponds to differential metabolic effects in the ovarian tumors from obese
Increased efficacy of metformin corresponds to differential metabolic effects in the ovarian tumors from obese
Oncotarget (2018-01-18)
Jianjun Han, Weiya Z Wysham, Yan Zhong, Hui Guo, Lu Zhang, Kim M Malloy, Hallum K Dickens, Gene Huh, Douglas Lee, Liza Makowski, Chunxiao Zhou, Victoria L Bae-Jump
PMID29340030
ABSTRACT
Obesity is a significant risk factor for ovarian cancer (OC) and associated with worse outcomes for this disease. We assessed the anti-tumorigenic effects of metformin in human OC cell lines and a genetically engineered mouse model of high grade serous OC under obese and lean conditions. Metformin potently inhibited growth in a dose-dependent manner in all four human OC cell lines through AMPK/mTOR pathways. Treatment with metformin resulted in G1 arrest, induction of apoptosis, reduction of invasion and decreased hTERT expression. In the K18-gT