2 x 12 pre-weighed scavengers / resins
Useful for:
Broad-spectrum metal removal
Impurity removal
Color treatment
Materials required – set up:
- 1 mylar bag from the KitAlysis Metal Scavenger Reaction Screening Kit and you will use the following components
- 2 x 12 pre-weighed scavengers / resins in glass vials topped with cap mat
Additional (not included) items needed:
- 1 (1 mL) syringe with needle or 1 (1 mL) pipette with tips
- 50 mL flask
- Solvent (DCM, THF, EtOAc, DMF, MeOH, Toluene, Water (pH 2-10) or any other solvent in which your substrate is soluble)
- Orbital shaker for 96 well plates (the use of stir bar is not recommended with some of the resins)
- Your substrate: metal contaminated crude or purified material
Loading options:
Each kit can be used to screen 12 different scavengers, each at high (50 mg/vial) and low (10 mg/vial) loadings. Stock solution guidance for various w/w% is provided below. For instance, in a single kit, both 100 w/w% and 20 w/w% scavenger loading can be screened with a given stock solution.