BR122578
BRAND® laboratory bottle, graduated with screw cap
capacity 20000 mL
別名:
瓶
スキップする
物質
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
ジョイント
Joints threaded neck (GL45)
無菌性
non-sterile
特徴
autoclavable
graduations
with cap
包装
pack of 1 ea
メーカー/製品名
BRAND 122578
キャパシティ
20000 mL
直径× H
300 mm × 510 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | BR122564 | BR122574 | BR122562 |
|---|---|---|---|
| feature autoclavable, with cap, graduations | feature autoclavable, graduations, with cap | feature autoclavable, graduations, with cap | feature autoclavable, graduations, with cap |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material borosilicate glass bottle, narrow-mouth bottle, round bottle, clear bottle, polypropylene cap | material borosilicate glass bottle, clear bottle, narrow-mouth bottle, polypropylene cap, round bottle | material borosilicate glass bottle, clear bottle, narrow-mouth bottle, polypropylene cap, round bottle | material borosilicate glass bottle, clear bottle, narrow-mouth bottle, polypropylene cap, round bottle |
| capacity 20000 mL | capacity 2000 mL | capacity 10000 mL | capacity 1000 mL |
| packaging pack of 1 ea | packaging pack of 10 ea | packaging pack of 1 ea | packaging pack of 10 ea |
| joint Joints threaded neck (GL45) | joint Joints threaded neck (GL45) | joint Joints threaded neck (GL 45) | joint Joints threaded neck (GL45) |