Merck
BR140300
BRAND® holder for separating funnel
for rods of 8 - 14 mm diam.
別名:
漏斗
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥3,289
¥3,289
出荷予定日2025年4月14日
サイズを選択してください
表示を変更する
1 EA
¥3,289
About This Item
UNSPSCコード:
41121809
NACRES:
NB.35
¥3,289
出荷予定日2025年4月14日
メーカー/製品名
BRAND 140300
詳細
The BRAND® Holder for Separating Funnel is designed to securely support one or two funnels with a top outer diameter ranging from 50 to 120 mm. Made from durable polypropylene (PP), this holder is built to withstand the demands of laboratory environments. It can be easily mounted on support rods with an outer diameter of 8-14 mm, featuring a locking nut for added stability and security. This versatile holder enhances the efficiency of your separation processes, providing reliable support for your funnels during experiments and ensuring safe and effective liquid handling.
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)