スキップする
BR150040
BRAND® watch glass
soda-lime glass, diam. 80 mm , ground edges stress-reduced
別名:
Dishes, Watch glasses
ログインで組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
サイズ/SKU在庫状況単価
10 ea
カートの在庫状況を確認する
¥8,790
この商品について
NACRES:
NB.43
UNSPSC Code:
41121801
eCl@ss:
32035490
Material:
soda-lime glass
¥8,790
カートの在庫状況を確認する
​
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
material
soda-lime glass
manufacturer/tradename
BRAND 150040
diam.
80 mm , ground edges stress-reduced
General description
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 1
当該品目
BR150050BR150060BR150065
BRAND® watch glass soda-lime glass, diam. 80 mm , ground edges stress-reduced
BR150040
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 100 mm , ground edges stress-reduced
BR150050
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 120 mm , ground edges stress-reduced
BR150060
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 125 mm , ground edges stress-reduced
BR150065
BRAND® watch glass
description
soda-lime glass, diam. 80 mm , ground edges stress-reduced
description
soda-lime glass, diam. 100 mm , ground edges stress-reduced
description
soda-lime glass, diam. 120 mm , ground edges stress-reduced
description
soda-lime glass, diam. 125 mm , ground edges stress-reduced
material
soda-lime glass
material
soda-lime glass
material
soda-lime glass
material
soda-lime glass
manufacturer/tradename
BRAND 150040
manufacturer/tradename
BRAND 150050
manufacturer/tradename
BRAND 150060
manufacturer/tradename
BRAND 150065
diam.
80 mm , ground edges stress-reduced
diam.
100 mm , ground edges stress-reduced
diam.
120 mm , ground edges stress-reduced
diam.
125 mm , ground edges stress-reduced
Still not finding the right product?
Explore all of our products under BRAND® watch glass
最新バージョンのいずれかを選択してください:
試験成績書(COA)