表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|---|---|
10 ea | カートの在庫状況を確認する | ¥13,700 |
この商品について
NACRES:
NB.43
UNSPSC Code:
41121801
Material:
soda-lime glass
¥13,700
カートの在庫状況を確認する
material
soda-lime glass
manufacturer/tradename
BRAND 150065
diam.
125 mm , ground edges stress-reduced
General description
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
当該品目 | |||
|---|---|---|---|
| description soda-lime glass, diam. 125 mm , ground edges stress-reduced | description soda-lime glass, diam. 40 mm , ground edges stress-reduced | description soda-lime glass, diam. 80 mm , ground edges stress-reduced | description soda-lime glass, diam. 90 mm , ground edges stress-reduced |
| material soda-lime glass | material soda-lime glass | material soda-lime glass | material soda-lime glass |
| diam. 125 mm , ground edges stress-reduced | diam. 40 mm , ground edges stress-reduced | diam. 80 mm , ground edges stress-reduced | diam. 90 mm , ground edges stress-reduced |
| manufacturer/tradename BRAND 150065 | manufacturer/tradename BRAND 150010 | manufacturer/tradename BRAND 150040 | manufacturer/tradename BRAND 150045 |
Still not finding the right product?
Explore all of our products under BRAND® watch glass