スキップする
BR150080
BRAND® watch glass
soda-lime glass, diam. 200 mm , ground edges stress-reduced
別名:
Dishes, Watch glasses
ログインで組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
あなたへ/カタログ番号在庫状況単価
この商品について
NACRES:
NB.43
UNSPSC Code:
41121801
Material:
soda-lime glass
現在、価格および在庫状況を閲覧できません。
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
material
soda-lime glass
manufacturer/tradename
BRAND 150080
diam.
200 mm , ground edges stress-reduced
General description
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 1
当該品目
BR150070BR150015BR150020
BRAND® watch glass soda-lime glass, diam. 200 mm , ground edges stress-reduced
BR150080
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 150 mm , ground edges stress-reduced
BR150070
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 50 mm , ground edges stress-reduced
BR150015
BRAND® watch glass
BRAND® watch glass soda-lime glass, diam. 60 mm , ground edges stress-reduced
BR150020
BRAND® watch glass
material
soda-lime glass
material
soda-lime glass
material
soda-lime glass
material
soda-lime glass
diam.
200 mm , ground edges stress-reduced
diam.
150 mm , ground edges stress-reduced
diam.
50 mm , ground edges stress-reduced
diam.
60 mm , ground edges stress-reduced
manufacturer/tradename
BRAND 150080
manufacturer/tradename
BRAND 150070
manufacturer/tradename
BRAND 150015
manufacturer/tradename
BRAND 150020
Still not finding the right product?
Explore all of our products under BRAND® watch glass
最新バージョンのいずれかを選択してください:
試験成績書(COA)