スキップする
BR150907
BRAND® watch glass
PTFE, diam. 65 mm
別名:
Dishes, PTFE, Watch glasses
ログインで組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
あなたへ/カタログ番号在庫状況単価
1 ea
カートの在庫状況を確認する
¥3,480
この商品について
NACRES:
NB.43
UNSPSC Code:
41121801
Material:
PTFE
¥3,480
カートの在庫状況を確認する
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
material
PTFE
manufacturer/tradename
BRAND 150907
diam.
65 mm
General description
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 1
当該品目
BR150904BR150910BR150922
BRAND® watch glass PTFE, diam. 65 mm
BR150907
BRAND® watch glass
BRAND® watch glass PTFE, diam. 40 mm
BR150904
BRAND® watch glass
BRAND® watch glass PTFE, diam. 75 mm
BR150910
BRAND® watch glass
BRAND® watch glass PTFE, diam. 150 mm
BR150922
BRAND® watch glass
description
PTFE, diam. 65 mm
description
PTFE, diam. 40 mm
description
PTFE, diam. 75 mm
description
PTFE, diam. 150 mm
material
PTFE
material
PTFE
material
PTFE
material
PTFE
diam.
65 mm
diam.
40 mm
diam.
75 mm
diam.
150 mm
manufacturer/tradename
BRAND 150907
manufacturer/tradename
BRAND 150904
manufacturer/tradename
BRAND 150910
manufacturer/tradename
BRAND 150922
Still not finding the right product?
Explore all of our products under BRAND® watch glass
最新バージョンのいずれかを選択してください:
試験成績書(COA)