1 EA
¥4,080
¥4,080
カートの在庫状況を確認する
この商品について
NACRES:
NB.43
UNSPSC Code:
41121801
スキップする
1 of 4
当該品目 | BR150922 | BR150907 | BR150900 |
|---|---|---|---|
| material PTFE | material PTFE | material PTFE | material PTFE |
| manufacturer/tradename BRAND 150910 | manufacturer/tradename BRAND 150922 | manufacturer/tradename BRAND 150907 | manufacturer/tradename BRAND 150900 |
| diam. 75 mm | diam. 150 mm | diam. 65 mm | diam. 20 mm |
General description
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG