Merck
BR156644
BRAND® tray stackable
size 370 x 310 x 75 mm
別名:
Photographoc trays, Trays
ログインで組織・契約価格をご覧ください。
サイズを選択してください
1 EA
¥5,250
¥5,250
カートの在庫状況を確認する
この商品について
NACRES:
NB.51
UNSPSC Code:
41122800
Manufacturer/tradename:
BRAND 156644
Material:
polypropylene , white
Packaging:
pack of 1 ea
スキップする
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
material
polypropylene , white
packaging
pack of 1 ea
manufacturer/tradename
BRAND 156644
size
370 x 310 x 75 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 4
当該品目
BR156642BR156646BR156640
BRAND® tray stackable size 370 x 310 x 75 mm
BR156644
BRAND® tray stackable
BRAND® tray stackable size 300 x 240 x 70 mm
BR156642
BRAND® tray stackable
BRAND® tray stackable size 510 x 410 x 120 mm
BR156646
BRAND® tray stackable
BRAND® tray stackable size 225 x 180 x 45 mm
BR156640
BRAND® tray stackable
material
polypropylene , white
material
polypropylene , white
material
polypropylene , white
material
polypropylene , white
manufacturer/tradename
BRAND 156644
manufacturer/tradename
BRAND 156642
manufacturer/tradename
BRAND 156646
manufacturer/tradename
BRAND 156640
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
size
370 x 310 x 75 mm
size
300 x 240 x 70 mm
size
510 x 410 x 120 mm
size
225 x 180 x 45 mm
General description
The BRAND® stackable tray is made from durable white polypropylene (PP), designed for versatile use in laboratory and industrial settings. Its ribbed bottom enhances stability, preventing contents from shifting during transport or storage. The reinforced rim adds strength, while the slightly sloping walls facilitate easy access and pouring. This stackable design allows for efficient storage, maximizing space when not in use. Ideal for organizing and handling various materials, this tray combines practicality with durability, making it an essential tool for any workspace.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)