Merck
All Photos(1)
BR23715
BRAND® spare burette for automatic burette Dr. Schilling
size 15 mL, clear
Synonym(s):
SILBERBRAND, ビュレット
NACRES:
NB.43
物質
AR-Glass
clear
詳細
SILBERBRAND
包装
pack of 1 ea
manufacturer/tradename
BRAND 23715
L
330 mm
サイズ
15 mL
Looking for similar products? Visit Product Comparison Guide
Compare Similar Items
View Full Comparison
1 of 4
This Item
BR23716BR23718BR23713
material
AR-Glass , clear
material
AR-Glass , clear
material
AR-Glass , clear
material
AR-Glass , clear
description
SILBERBRAND
description
SILBERBRAND
description
SILBERBRAND
description
SILBERBRAND
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23715
manufacturer/tradename
BRAND 23716
manufacturer/tradename
BRAND 23718
manufacturer/tradename
BRAND 23713
L
330 mm
L
380 mm
L
580 mm
L
330 mm
size
15 mL
size
25 mL
size
50 mL
size
10 mL
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service