BR23718
AR-Glass
clear
SILBERBRAND
pack of 1 ea
BRAND 23718
580 mm
50 mL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | BR23716 | BR23715 | BR23713 |
---|---|---|---|
material AR-Glass , clear | material AR-Glass , clear | material AR-Glass , clear | material AR-Glass , clear |
description SILBERBRAND | description SILBERBRAND | description SILBERBRAND | description SILBERBRAND |
packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
manufacturer/tradename BRAND 23718 | manufacturer/tradename BRAND 23716 | manufacturer/tradename BRAND 23715 | manufacturer/tradename BRAND 23713 |
L 580 mm | L 380 mm | L 330 mm | L 330 mm |
size 50 mL | size 25 mL | size 15 mL | size 10 mL |
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
How to Find the Product Number
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
How to Find a Lot/Batch Number for COA
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service