表示を変更する
| 容量 | カタログ番号 | 在庫状況 | 単価 |
|---|---|---|---|
| 2 ea | カートの在庫状況を確認する | ¥8,550 |
この商品について
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
2 mL
Material:
polypropylene stopper, transparent borosilicate glass 3.3 flask
Packaging:
pack of 2 ea
¥8,550
カートの在庫状況を確認する
material
polypropylene stopper, transparent borosilicate glass 3.3 flask
joint
joint: ST/NS7/16
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36402
capacity
2 mL
General description
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
当該品目 | |||
|---|---|---|---|
| material polypropylene stopper, transparent borosilicate glass 3.3 flask | material polypropylene stopper, transparent borosilicate glass 3.3 flask | material polypropylene stopper, transparent borosilicate glass 3.3 flask | material polypropylene stopper, transparent borosilicate glass 3.3 flask |
| capacity 2 mL | capacity 1 mL | capacity 20 mL | capacity 10 mL |
| feature accuracy class A | feature accuracy class A | feature accuracy class A | feature accuracy class A |
| manufacturer/tradename BRAND 36402 | manufacturer/tradename BRAND 36401 | manufacturer/tradename BRAND 36406 | manufacturer/tradename BRAND 36404 |
| joint joint: ST/NS7/16 | joint joint ST/NS neck7/16 | joint joint: ST/NS10/19 | joint joint: ST/NS10/19 |
| packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea |
グローバルトレードアイテム番号
| カタログ番号 | GTIN |
|---|---|
| BR36402-2EA | 04061831865264 |