Merck
All Photos(1)
BR463036
BRAND® rubber conical gasket for filter funnels and filter flasks
size 63 mm
Synonym(s):
Filter, Rubber gasket
NACRES:
NB.24
物質
EPDM rubber
包装
pack of 10 ea
manufacturer/tradename
BRAND 463036
サイズ
63 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Compare Similar Items
View Full Comparison
1 of 4
This Item
BR462827BR462932BR463139
material
EPDM rubber
material
EPDM rubber
material
EPDM rubber
material
EPDM rubber
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
manufacturer/tradename
BRAND 463036
manufacturer/tradename
BRAND 462827
manufacturer/tradename
BRAND 462932
manufacturer/tradename
BRAND 463139
size
63 mm
size
42 mm
size
50 mm
size
76 mm
詳細
Conical gaskets to place between filter funnel and filter flask.
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service