Merck
BR705820
BRAND® Transferpette® S Pipette
tip volume (25 μL), single-channel, fixed volume, mechanical, DE-M
ログインto View Organizational & Contract Pricing
特徴
single-channel fixed volume
mechanical
包装
case of 1 ea
manufacturer/tradename
BRAND 705820
チップ容量
(25 μL)
類似した製品をお探しですか? Visit 製品比較ガイド
関連するカテゴリー
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 4
当該品目
BR705862BR705828BR705844
BRAND® Transferpette® S Pipette tip volume (25 μL), single-channel, fixed volume, mechanical, DE-M
BR705820
BRAND® Transferpette® S Pipette
BRAND® Transferpette® S Pipette tip volume (1000 μL), single-channel, fixed volume, mechanical, DE-M
BR705862
BRAND® Transferpette® S Pipette
BRAND® Transferpette® S Pipette tip volume (50 μL), single-channel, fixed volume, mechanical, DE-M
BR705828
BRAND® Transferpette® S Pipette
BRAND® Transferpette® S Pipette tip volume (200 μL), single-channel, fixed volume, mechanical, DE-M
BR705844
BRAND® Transferpette® S Pipette
packaging
case of 1 ea
packaging
case of 1 ea
packaging
case of 1 ea
packaging
case of 1 ea
manufacturer/tradename
BRAND 705820
manufacturer/tradename
BRAND 705862
manufacturer/tradename
BRAND 705828
manufacturer/tradename
BRAND 705844
tip volume
(25 μL)
tip volume
(1000 μL)
tip volume
(50 μL)
tip volume
(200 μL)
詳細
The Transferpette S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
Overview- Volume range of single-channel pipette: from 0.1 μl to 10 ml- Volume range of multi-channel pipette: from 0.5 μl to 300 μl- Short stroke and low operating forces- Universal tip cone- Completely autoclaving at 121 μC- One-hand operation for right- and left-handed users- Color code on volume adjustment wheel, shaft and volume-change protection- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes- Can be calibrated easily and without tools thanks to Easy Calibration technology- Slim shaft for sampling from narrow vessels (centrifuge tubes)- Operating temperature from +15 μC to +40 μC- Vapor pressure up to 500 mbar- Viscosity up to 260 mPa s
Efficient: One-hand operation
Flexible: ergonomic design for all hand sizes
Reliable: volume-change protection prevents accidental adjustments
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
試験成績書(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください (: T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
以下のように入力してください 1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
ロット番号とバッチ番号は、製品ラベルで「Lot」または「Batch」の言葉に続いて記載されています。
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
テクニカルサービス連絡先