Merck
All Photos(2)
BR780505
BRAND® disposable microcentrifuge tube
without lid, capacity 1.5 mL
Synonym(s):
チューブ, マイクロ遠心チューブ
物質
polypropylene
無菌性
non-sterile
特徴
without lid
包装
pack of 2000
manufacturer/tradename
BRAND 780505
パラメーター
6000 × g max. RCF
キャパシティ
1.5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Compare Similar Items
View Full Comparison
1 of 4
This Item
BR780540Z640921Z640883
BRAND® centrifuge tubes capacity 75 mL
Z640921
BRAND® centrifuge tubes
BRAND® centrifuge tubes capacity 10 mL
Z640883
BRAND® centrifuge tubes
material
polypropylene
material
clear polypropylene
material
polypropylene , round bottom tube
material
polypropylene , round bottom tube
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
feature
without lid
feature
with safety lid
feature
autoclavable at 121°C and withstand RCF of up to 5,000 × g
feature
autoclavable at 121°C and withstand RCF of up to 5,000 × g
packaging
pack of 2000
packaging
pack of 1000
packaging
pack of 20
packaging
pack of 250
manufacturer/tradename
BRAND 780505
manufacturer/tradename
BRAND 780540
manufacturer/tradename
BRAND 115352
manufacturer/tradename
BRAND 115342
parameter
6000 × g max. RCF
parameter
30000 × g max. RCF
parameter
-
parameter
-
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service