Merck
BR82220
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Straight, for burette capacity 2-10 mL
別名:
ビュレット
NACRES:
NB.43
物質
polystyrene
特徴
for burette capacity 2-10 mL
包装
pack of 1 ea
manufacturer/tradename
BRAND 82220
類似した製品をお探しですか? Visit 製品比較ガイド
関連するカテゴリー
類似品目との比較
比較内容をすべて表示
Show Differences
1 of 4
当該品目
BR82222BR82121BR82170
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
feature
for burette capacity 2-10 mL
feature
for burette capacity 50 mL
feature
for burette capacity 25 mL
feature
for burette capacity 2-50 mL
manufacturer/tradename
BRAND 82220
manufacturer/tradename
BRAND 82222
manufacturer/tradename
BRAND 82121
manufacturer/tradename
BRAND 82170
詳細
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください ( T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
テクニカルサービス連絡先