CDS020910

Sigma-Aldrich

4-Methoxy-trans-β-styrylboronic acid pinacol ester

Empirical Formula (Hill Notation):
C15H21BO3
分子量:
260.14
MDL番号:
PubChem Substance ID:

詳細

AldrichCPR

SMILES string

CC1(C)OB(/C=C/C(C=C2)=CC=C2OC)OC(C)1C

InChI

1S/C15H21BO3/c1-14(2)15(3,4)19-16(18-14)11-10-12-6-8-13(17-5)9-7-12/h6-11H,1-5H3/b11-10+

InChI key

VXSHTICWQKLRMP-ZHACJKMWSA-N

その他情報

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

危険有害性情報

hazcat

Aquatic Chronic 4

storage_class_code

13 - Non Combustible Solids

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)

原産地証明書(COO)

その他のドキュメント

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先