物質
pennyhead glass cap (stopper)
包装
pkg of 24 ea
メーカー/製品名
WHEATON® 227497-03G
ボトル 直径× H
69 mm × 165 mm
キャパシティ
300 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
特徴および利点
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
法的情報
WHEATON is a registered trademark of Wheaton Industries, Inc.
最新バージョンのいずれかを選択してください:
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)