DWK233301504
glass
non-sterile
graduation: no
pack of 250 ea
pkg of 4 packs
DURAN®
(2 mL)
1 of 4
当該品目 | DWK233302303 | Z740086 | Z740085 |
---|---|---|---|
capacity (2 mL) | capacity (2 mL) | capacity - | capacity - |
sterility non-sterile | sterility non-sterile | sterility non-sterile; autoclaved | sterility non-sterile; autoclaved |
feature graduation: no | feature graduation: no | feature bulk, XXL | feature bulk |
packaging pack of 250 ea | packaging pack of 250 ea, pkg of 4 packs | packaging pack of 1000 ea (5 bags, 200 each) | packaging pack of 200 ea (1 bag of 200) |
manufacturer/tradename DURAN® | manufacturer/tradename DURAN® | manufacturer/tradename BRAND 702600 | manufacturer/tradename BRAND 702595 |
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
製品番号の調べ方
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください (例 T1503).
例
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
COAのロット番号・バッチ番号の調べ方
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
テクニカルサービス連絡先