Merck
All Photos(1)
DWK34507-99
KIMBLE® KIMAX® open ended melting capillary
glass, L (100 mm)
物質
glass
translucent borosilicate glass tube
無菌性
non-sterile
包装
case of 20 ea 100 tubes in each vial
manufacturer/tradename
Kimble® 34507-99
L
(100 mm)
チューブ直径
(1.5-1.8 mm)
Looking for similar products? Visit Product Comparison Guide
Compare Similar Items
View Full Comparison
1 of 4
This Item
DWK34500-99DWK34502-99KIM3450599
material
glass
material
glass , translucent borosilicate glass tube
material
glass , translucent borosilicate glass tube
material
borosilicate glass
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
-
packaging
case of 20 ea 100 tubes in each vial
packaging
case of 20 ea 100 tubes in each vial
packaging
case of 20 ea 100 tubes in each vial
packaging
case of 20 × 100 ea (2000 total)
manufacturer/tradename
Kimble® 34507-99
manufacturer/tradename
Kimble® 34500-99
manufacturer/tradename
Kimble® 34502-99
manufacturer/tradename
-
L
(100 mm)
L
(100 mm)
L
(100 mm)
L
-
tube diam.
(1.5-1.8 mm)
tube diam.
(1.5-1.8 mm)
tube diam.
(1.5-1.8 mm)
tube diam.
-
詳細
Melting Point Capillary
  • KIMAX melting point capillary tubes are used to determine the point at which a solid sample turns to liquid.
  • Tubes are 100 mm long when supplied with both ends open, so that approximately 90 mm will remain after sealing
  • Tube length insures that the open end will extend well above the bath′s surface to prevent the bath liquid from entering the tube
  • 34502-99 and 34507-99 have a smaller I.D. than 34500-99 and are designed from requirements contained in the United States Pharmacopeia (USP)
  • Packaged with 100 tubes per vial and 20 vials per case
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
法的情報
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service