Merck
All Photos(1)

DWK63C50

Kimble® Disposable Pasteur Pipet

L 5.75 in.

包装

pkg of 1000 ea

manufacturer/tradename

Kimble® 63C50

L

5.75 in.

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
DWK63B93DWK63A54PDWK63DP1005
packaging

pkg of 1000 ea

packaging

pkg of 1000 ea

packaging

pkg of 1000 ea

packaging

pkg of 800 ea

manufacturer/tradename

Kimble® 63C50

manufacturer/tradename

Kimble® 63B93

manufacturer/tradename

Kimble® 63A54P

manufacturer/tradename

Kimble® 63DP1005

L

5.75 in.

L

9.0 in.

L

5.75 in.

L

5.75 in.

アプリケーション

Bacteriology, immunology, hematology, and serology

特徴および利点

Removes risk of cross-contamination of high value specimens and laboratory reagents.

法的情報

KIMBLE is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service