DWK73750-16100
KIMBLE® disposable screw thread culture tube with white marking spot
round bottom borosilicate glass, tube diam. × L (16 mm) (100 mm), non-sterile
サイズを選択してください
¥89,000
サイズを選択してください
About This Item
¥89,000
物質
round bottom borosilicate glass
round bottom
screw top neck
transparent borosilicate glass tube
無菌性
non-sterile
特徴
without cap
包装
pkg of 1000 ea
メーカー/製品名
Kimble® 73750-16100
チューブ直径× L
(16 mm) (100 mm)
チューブ直径
(16 mm)
フィッティング
neck (External Thread)
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | DWK73750-13100 | DWK73750-16125 | DWK73750-16150 |
---|---|---|---|
material round bottom borosilicate glass , screw top neck, transparent borosilicate glass tube | material round bottom borosilicate glass , screw top neck, transparent borosilicate glass tube | material round bottom borosilicate glass , screw top neck, transparent borosilicate glass tube | material round bottom borosilicate glass , screw top neck, transparent borosilicate glass tube |
sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
feature without cap | feature without cap | feature without cap | feature without cap |
packaging pkg of 1000 ea | packaging case of 1000 ea Modular tray shrink-wrapped in plastic film | packaging case of 1000 ea Modular tray shrink-wrapped in plastic film | packaging case of 1000 ea Modular tray shrink-wrapped in plastic film |
manufacturer/tradename Kimble® 73750-16100 | manufacturer/tradename Kimble® 73750-13100 | manufacturer/tradename Kimble® 73750-16125 | manufacturer/tradename Kimble® 73750-16150 |
tube diam. × L (16 mm) (100 mm) | tube diam. × L (13 mm) (100 mm) | tube diam. × L (16 mm) (125 mm) | tube diam. × L (16 mm) (105 mm) |
詳細
- These culture tubes are suitable for tissue culture work and general bacteriological use.
- Round-bottomed
- Each tube has a chevron marking area
- Supplied without caps
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
特徴および利点
- Manufactured from borosilicate glass 5.1
- Round-bottomed
- Each tube has a chevron marking area
- Modular trays are shrink-wrapped in plastic film
- Supplied without caps
法的情報
最新バージョンのいずれかを選択してください:
アクティブなフィルタ