DWK73785-10
KIMBLE® centrifuge tube
with screw-top finish, conical bottom clear glass, capacity (10 mL), without cap, non-sterile
サイズを選択してください
¥31,000
サイズを選択してください
About This Item
¥31,000
物質
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
無菌性
non-sterile
特徴
without cap
包装
case of 125 ea Modular trays are shrink-wrapped
メーカー/製品名
Kimble® 73785-10
キャパシティ
(10 mL)
フィッティング
15-415 neck for thread
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | DWK73785-15 | DWK73785-50 | DWK73790-5 |
---|---|---|---|
material conical bottom clear glass , transparent borosilicate glass tube | material conical bottom clear glass , transparent borosilicate glass tube | material conical bottom clear glass , transparent borosilicate glass tube | material conical bottom clear glass , transparent borosilicate glass tube |
sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
feature without cap | feature without cap | feature without cap | feature - |
packaging case of 125 ea Modular trays are shrink-wrapped | packaging case of 125 ea Modular trays are shrink-wrapped | packaging case of 72 ea Modular trays are shrink-wrapped | packaging case of 125 ea shrink wrapped in modular tray |
manufacturer/tradename Kimble® 73785-10 | manufacturer/tradename Kimble® 73785-15 | manufacturer/tradename Kimble® 73785-50 | manufacturer/tradename Kimble® 73790-5 |
capacity (10 mL) | capacity (15 mL) | capacity (50 mL) | capacity (5 mL) |
詳細
- These tubes are suitable for tissue culture work and general bacteriological use.
- Supplied without caps
- No marking spot
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
法的情報
最新バージョンのいずれかを選択してください:
アクティブなフィルタ