Merck
All Photos(1)
DWK73790-50
KIMBLE® centrifuge tube
with plain rim finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
物質
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
無菌性
non-sterile
包装
case of 72 ea shrink wrapped in modular tray
manufacturer/tradename
Kimble® 73790-50
キャパシティ
(50 mL)
フィッティング
neck (Pushfit)
Looking for similar products? Visit Product Comparison Guide
Compare Similar Items
View Full Comparison
1 of 4
This Item
DWK73790-10DWK73790-5DWK73790-15
KIMBLE® centrifuge tube with plain rim finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
DWK73790-50
KIMBLE® centrifuge tube
KIMBLE® centrifuge tube with plain rim finish, conical bottom clear glass, capacity (10 mL), without cap, non-sterile
DWK73790-10
KIMBLE® centrifuge tube
KIMBLE® centrifuge tube with plain rim finish, conical bottom clear glass, capacity (5 mL), without cap, non-sterile
DWK73790-5
KIMBLE® centrifuge tube
KIMBLE® centrifuge tube with plain rim finish, conical bottom clear glass, capacity (15 mL), without cap, non-sterile
DWK73790-15
KIMBLE® centrifuge tube
material
conical bottom clear glass , transparent borosilicate glass tube
material
conical bottom clear glass , transparent borosilicate glass tube
material
conical bottom clear glass , transparent borosilicate glass tube
material
V-bottom bottom, conical bottom clear glass , transparent borosilicate glass tube
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
packaging
case of 72 ea shrink wrapped in modular tray
packaging
case of 125 ea shrink wrapped in modular tray
packaging
case of 125 ea shrink wrapped in modular tray
packaging
case of 125 ea shrink wrapped in modular tray
manufacturer/tradename
Kimble® 73790-50
manufacturer/tradename
Kimble® 73790-10
manufacturer/tradename
Kimble® 73790-5
manufacturer/tradename
Kimble® 73790-15
capacity
(50 mL)
capacity
(10 mL)
capacity
(5 mL)
capacity
(15 mL)
fitting
neck (Pushfit)
fitting
neck (Pushfit)
fitting
neck (Pushfit)
fitting
neck (Pushfit)
詳細
Plain, disposable centrifuge tube can withstand centrifugation up to 2,980 RCF. Closures are not supplied
  • Conical bottom
  • The 5, 10, 15 sizes have a finished top for a snap-cap closure (not supplied)
  • The 50mL size has a tooled top
  • Shrink wrapped in modular trays to keep them clean and safe in transmit
  • Manufactured from 51 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class B requirements
Screw Thread Disposable Centrifuge Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
法的情報
KIMBLE is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service