Merck
DWK90P20150F
KIMBLE® disposable screw thread culture tube
flat bottom borosilicate glass, tube diam. × L (20 mm) (150 mm), non-sterile
物質
flat bottom borosilicate glass
flat bottom
screw top neck
transparent borosilicate glass tube
無菌性
non-sterile
特徴
without cap
包装
case of 500 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 90P20150F
チューブ直径× L
(20 mm) (150 mm)
フィッティング
neck (External Thread)
類似した製品をお探しですか? Visit 製品比較ガイド
類似品目との比較
比較内容をすべて表示
Show Differences
1 of 4
当該品目
DWK90K16125FDWK90H13100FDWK90K16100F
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
feature
without cap
feature
without cap
feature
without cap
feature
without cap
packaging
case of 500 ea Modular tray shrink-wrapped in plastic film
packaging
case of 1000 ea Modular tray shrink-wrapped in plastic film
packaging
case of 1000 ea Modular tray shrink-wrapped in plastic film
packaging
case of 1000 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 90P20150F
manufacturer/tradename
Kimble® 90K16125F
manufacturer/tradename
Kimble® 90H13100F
manufacturer/tradename
Kimble® 90K16100F
tube diam. × L
(20 mm) (150 mm)
tube diam. × L
(16 mm) (125 mm)
tube diam. × L
(13 mm) (100 mm)
tube diam. × L
(16 mm) (100 mm)
詳細
Disposable Screw Thread Culture Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
法的情報
KIMBLE is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください ( T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
テクニカルサービス連絡先