DWK92812G-5
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with glass stopper
glass flask, flask capacity (5 mL), class A
ログイン組織・契約価格を表示する
サイズを選択してください
6 EA
¥40,900
¥40,900
カートの在庫状況を確認する
サイズを選択してください
表示を変更する
6 EA
¥40,900
About This Item
UNSPSCコード:
41121804
NACRES:
NB.43
¥40,900
カートの在庫状況を確認する
物質
glass flask
glass stopper
特徴
class A
包装
case of 6 ea
メーカー/製品名
Kimble® 92812G-5
フラスコ容量
(5 mL)
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | DWK92812G-50 | DWK92812G-200 | DWK92812G-500 |
---|---|---|---|
material glass flask, glass stopper | material glass flask, glass stopper | material borosilicate glass flask, glass stopper | material glass flask, glass stopper |
manufacturer/tradename Kimble® 92812G-5 | manufacturer/tradename Kimble® 92812G-50 | manufacturer/tradename Kimble® 92812G-200 | manufacturer/tradename Kimble® 92812G-500 |
packaging case of 6 ea | packaging case of 6 ea | packaging case of 6 ea | packaging case of 6 ea |
feature class A | feature class A | feature class A | feature class A |
flask capacity (5 mL) | flask capacity (50 mL) | flask capacity (200 mL) | flask capacity - |
詳細
Class A Heavy Duty Wide Mouth Volumetric Flasks with Glass Stopper
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
特徴および利点
- Manufactured from borosilicate glass 3.3
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Heavy duty walls provide superior strength, durability and safety
- Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Printed large volume numbers allow quick identification when selecting glassware
- Flat interior bottoms for stirring with standard stir bars
- Calibrate to contain
- Graduated to Class A volumetric tolerances
- Supplied with or without stopper
法的情報
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
最新バージョンのいずれかを選択してください:
アクティブなフィルタ