表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥20,400 |
この商品について
NACRES:
NB.51
UNSPSC Code:
41121809
Joint:
joint: ST/NS19/22
Packaging:
pkg of 1 ea
¥20,400
カートの在庫状況を確認する
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F459025
capacity
25 mL , 25 mL
General description
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| capacity 25 mL , 25 mL | capacity 25 mL , 25 mL | capacity 25 mL , 25 mL | capacity 250 mL , 250 mL |
| manufacturer/tradename Synthware F459025 | manufacturer/tradename Synthware F451025 | manufacturer/tradename Synthware F451025A | manufacturer/tradename Synthware F451250A |
| joint joint: ST/NS19/22 | joint joint: ST/NS14/20 | joint joint: ST/NS14/20 | joint joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper