表示を変更する
| サイズ/SKU | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥12,600 |
この商品について
NACRES:
NB.51
UNSPSC Code:
41121809
Joint:
Joints: (16)
Packaging:
pkg of 1 ea
¥12,600
カートの在庫状況を確認する
joint
Joints: (16)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F471660
capacity
60 mL , 60 mL
General description
Supplied with a Glass or PTFE stopper
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| description 60 mL, Joints: (16) | description 60 mL, Joints: (16) | description 50 mL, top joint: ST/NS 14/20 | description 30 mL, Joints: (13) |
| manufacturer/tradename Synthware F471660 | manufacturer/tradename Synthware F471660A | manufacturer/tradename Synthware F471050 | manufacturer/tradename Synthware F471330A |
| capacity 60 mL , 60 mL | capacity 60 mL, 60 mL | capacity 50 mL, 50 mL | capacity 30 mL, 30 mL |
| joint Joints: (16) | joint Joints: (16) | joint joint: ST/NS top14/20 | joint Joints: (13) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Squibb-style separatory funnel with PTFE stopcock and PTFE stopper