表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥27,100 |
この商品について
NACRES:
NB.51
UNSPSC Code:
41121809
Joint:
joint: ST/NS24/40
Packaging:
pkg of 1 ea
¥27,100
カートの在庫状況を確認する
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F474000
capacity
1000 mL , 1000 mL
General description
Supplied with a Glass or PTFE stopper
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| description 1000 mL, joint: ST/NS 24/40 | description 1000 mL, top joint: ST/NS 24/40 | description 1000 mL, Joints: (27) | description 1000 mL, Joints: (27) |
| capacity 1000 mL , 1000 mL | capacity 1000 mL , 1000 mL | capacity 1000 mL , 1000 mL | capacity 1000 mL , 1000 mL |
| manufacturer/tradename Synthware F474000 | manufacturer/tradename Synthware F474000A | manufacturer/tradename Synthware F477000 | manufacturer/tradename Synthware F477000A |
| joint joint: ST/NS24/40 | joint joint: ST/NS top24/40 | joint Joints: (27) | joint Joints: (27) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Squibb-style separatory funnel with PTFE stopcock and PTFE stopper