SYNP170001
Synthware™ round bottom pressure vessel with PTFE bushing
75 mL, O.D. × L 46 mm × 129 mm, #15 bushing
別名:
PTFE Bushing Vessel, Pressure Vessel with Bushing, Round Bottom Pressure Vessel, Synthware P170001, pressure tube
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥12,990
¥12,990
カートの在庫状況を確認する
すべての画像(1)
サイズを選択してください
表示を変更する
1 EA
¥12,990
About This Item
UNSPSCコード:
41100000
NACRES:
NB.51
¥12,990
カートの在庫状況を確認する
おすすめの製品
1 of 4
当該品目 | SYNP170003 | SYNP170002 | SYNP170004 |
---|---|---|---|
capacity 75 mL , 75 mL | capacity 150 mL , 150 mL | capacity - | capacity - |
packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
manufacturer/tradename Synthware P170001 | manufacturer/tradename Synthware P170003 | manufacturer/tradename Synthware P170002 | manufacturer/tradename Synthware P170004 |
O.D. × L 46 mm × 129 mm | O.D. × L 60 mm × 139 mm | O.D. × L 50 mm × 137 mm | O.D. × L 60 mm × 147 mm |
fitting #15 bushing | fitting #15 bushing | fitting - | fitting - |
詳細
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
法的情報
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
最新バージョンのいずれかを選択してください:
この製品を見ている人はこちらもチェック