Merck
SYNP170003
Synthware round bottom pressure vessel with PTFE bushing
150 mL, O.D. × L 60 mm × 139 mm, #15 bushing
別名:
Pressure Vessel, Synthware P170003, pressure tube
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥13,510
¥13,510
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥13,510
About This Item
UNSPSCコード:
41100000
NACRES:
NB.51
¥13,510
Check Cart for Availability
包装
pkg of 1 ea
メーカー/製品名
Synthware P170003
外径 × L
60 mm × 139 mm
キャパシティ
150 mL
150 mL
フィッティング
#15 bushing
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
法的情報
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
最新バージョンのいずれかを選択してください:
試験成績書(COA)