It is a good equipment of glass regarding its quality however I could not find and data related to pressure rate so I do not how far I can go with temperature. Solvent is water.
SYNP170005
Synthware™ round bottom pressure vessel with PTFE bushing
350 mL, O.D. × L 80 mm × 154 mm, #15 bushing
Synthware™ round bottom pressure vessel with PTFE bushing
別名:
Pressure Vessel, Synthware P170005, pressure tube
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥15,800
¥15,800
カートの在庫状況を確認する
サイズを選択してください
表示を変更する
1 EA
¥15,800
About This Item
UNSPSCコード:
41100000
NACRES:
NB.51
¥15,800
カートの在庫状況を確認する
包装
pkg of 1 ea
メーカー/製品名
Synthware P170005
外径 × L
80 mm × 154 mm
キャパシティ
350 mL
フィッティング
#15 bushing
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | SYNP170002 | SYNP170004 | SYNP170006 |
---|---|---|---|
capacity 350 mL | capacity 75 mL | capacity 150 mL | capacity 350 mL |
packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
manufacturer/tradename Synthware P170005 | manufacturer/tradename Synthware P170002 | manufacturer/tradename Synthware P170004 | manufacturer/tradename Synthware P170006 |
O.D. × L 80 mm × 154 mm | O.D. × L 50 mm × 137 mm | O.D. × L 60 mm × 147 mm | O.D. × L 80 mm × 162 mm |
fitting #15 bushing | fitting - | fitting - | fitting - |
詳細
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
法的情報
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
最新バージョンのいずれかを選択してください:
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)