¥19,490
カートの在庫状況を確認する
おすすめの製品
1 of 4
当該品目 | SYNP170002 | SYNP170004 | SYNP170005 |
---|---|---|---|
manufacturer/tradename Synthware P170006 | manufacturer/tradename Synthware P170002 | manufacturer/tradename Synthware P170004 | manufacturer/tradename Synthware P170005 |
capacity 350 mL | capacity 75 mL | capacity 150 mL | capacity 350 mL |
packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
O.D. × L 80 mm × 162 mm | O.D. × L 50 mm × 137 mm | O.D. × L 60 mm × 147 mm | O.D. × L 80 mm × 154 mm |
詳細
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
法的情報
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
最新バージョンのいずれかを選択してください:
この製品を見ている人はこちらもチェック