Product number Z114278, plier-type decapper, is autoclavable (steam only) up to 121 Degrees C.
O.D.
20 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | |||
|---|---|---|---|
| O.D. 20 mm | O.D. 11 mm | O.D. - | O.D. - |
General description
Removes aluminum seals from serum bottles and vials.
related product
製品番号
詳細
価格
-
Is product number Z114278, plier-type decapper, autoclavable?
1 回答-
役に立ちましたか?
-
-
What is the Department of Transportation shipping information for this product?
1 回答-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.