Merck
Z130931
Nalgene® Griffin beakers
capacity 50 mL, translucent polypropylene
ログインto View Organizational & Contract Pricing
Select a Size
別名:
PP beaker, nalgene PP beaker
NACRES:
NB.43
物質
translucent polypropylene
特徴
graduations
包装
pkg of 12 ea
manufacturer/tradename
Nalgene® 1201
キャパシティ
50 mL
類似した製品をお探しですか? Visit 製品比較ガイド
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 4
当該品目
Z130958Z130974Z130907
Nalgene® Griffin beakers capacity 50 mL, translucent polypropylene
Z130931
Nalgene® Griffin beakers
Nalgene® Griffin beakers capacity 100 mL, translucent polypropylene
Z130958
Nalgene® Griffin beakers
Nalgene® Griffin beakers capacity 250 mL, translucent polypropylene
Z130974
Nalgene® Griffin beakers
Nalgene® Griffin beakers capacity 150 mL, transparent polymethylpentene
Z130907
Nalgene® Griffin beakers
feature
graduations
feature
graduations
feature
graduations
feature
graduations
capacity
50 mL
capacity
100 mL
capacity
250 mL
capacity
150 mL
packaging
pkg of 12 ea
packaging
pkg of 12 ea
packaging
pkg of 6 ea
packaging
pkg of 12 ea
manufacturer/tradename
Nalgene® 1201
manufacturer/tradename
Nalgene® 1201
manufacturer/tradename
Nalgene® 1201
manufacturer/tradename
Nalgene® 1203
詳細
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
その他情報
Browse all plastic beakers
法的情報
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください (: T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
以下のように入力してください 1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
ロット番号とバッチ番号は、製品ラベルで「Lot」または「Batch」の言葉に続いて記載されています。
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
Aaron F Cipriano et al.
Acta biomaterialia, 62, 397-417 (2017-08-19)
This article reports anodization of Mg in KOH electrolyte and the associated surface, degradation, and biological properties for bioresorbable implant applications. The preparation procedures for electrodes and anodization setup significantly enhanced reproducibility of samples. The results of anodization performed at
テクニカルサービス連絡先